Search
Search

PREX1| Gene Symbol: PREX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028828497.1 | XP_028684330.1 | 3759 | 59 | ||
| GenBank mRNA | JU476548.1 | - | 3633 | 59 | ||
| JV657163.1 | - | 3640 | 59 | |||
Set Membership: |
|