Search
Search

CAPN13| Gene Symbol: CAPN13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112042.2 | XP_014967528.2 | 1989 | 91 | ||
| XM_015112043.2 | XP_014967529.2 | 1989 | 91 | |||
| XM_028831804.1 | XP_028687637.1 | 1989 | 91 | |||
Set Membership: |
|