Search
Search

KCNJ15| Gene Symbol: KCNJ15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132901.2 | XP_014988387.1 | 1874 | 88 | ||
| XM_015132902.2 | XP_014988388.1 | 345 | 88 | |||
| XM_015132903.2 | XP_014988389.1 | 124 | 88 | |||
Set Membership: |
|