Search
Search
| Gene Symbol: Hspb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053612.2 | NP_446064.1 | 1 - 2 | 725 | 65 | |
| GenBank mRNA | AF314540.1 | - | 1 - 2 | 407 | 65 | |
| BC061748.1 | - | 1 - 2 | 725 | 5599613 | 65 | |
Set Membership: |
|