Search
Search

Chrna10| Gene Symbol: Chrna10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022639.1 | NP_072161.1 | 3 - 4 | 422 | 85 | |
| XM_006229891.2 | XP_006229953.1 | 2 - 3 | 513 | 85 | ||
| GenBank mRNA | AF196344.1 | - | 3 - 4 | 422 | 85 | |
Set Membership: |
|