Search
Search
| Gene Symbol: Rgs19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021661.2 | NP_067693.1 | 4 - 5 | 522 | 78 | |
| XM_006235801.2 | XP_006235863.1 | 4 - 5 | 3283 | 78 | ||
| XM_008762540.1 | XP_008760762.1 | 3 - 4 | 795 | 78 | ||
| XM_008762541.1 | XP_008760763.1 | 3 - 4 | 262 | 78 | ||
| XM_008762542.1 | XP_008760764.1 | 3 - 4 | 258 | 78 | ||
| GenBank mRNA | AF068136.1 | - | 3 - 4 | 230 | 78 | |
| BC088141.1 | - | 4 - 5 | 522 | 7377022 | 78 | |
Set Membership: |
|