Search
Search
| Gene Symbol: Pccb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017030.2 | NP_058726.2 | 6 - 7 | 664 | 63 | |
| GenBank mRNA | BC078855.1 | - | 6 - 7 | 664 | 7108495 | 63 |
| M14634.1 | - | 6 - 7 | 665 | 63 | ||
Set Membership: |
|