Search
Search
| Gene Symbol: Pgm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108772.1 | NP_001102242.1 | 3 - 4 | 501 | 83 | |
| XM_006243482.2 | XP_006243544.1 | 3 - 4 | 513 | 83 | ||
| XM_008766451.1 | XP_008764673.1 | 3 - 4 | 529 | 83 | ||
| GenBank mRNA | BC166838.1 | - | 3 - 4 | 423 | 9027278 | 83 |
Set Membership: |
|