Search
Search

Cyp3a62| Gene Symbol: Cyp3a62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024232.1 | NP_001019403.1 | 7 - 8 | 883 | 132 | |
| GenBank mRNA | AB084894.1 | - | 7 - 8 | 883 | 132 | |
Set Membership: |
|