Search
Search
| Gene Symbol: Tceal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_577026.6 | XP_577026.4 | 1 - 2 | 166 | 81 | |
| GenBank mRNA | FQ213674.1 | - | 1 - 2 | 143 | 81 | |
| FQ213749.1 | - | 1 - 2 | 104 | 81 | ||
Set Membership: |
|