Search
Search
| Gene Symbol: Commd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108762.1 | NP_001102232.1 | 1 - 2 | 28 | 57 | |
| XM_006243133.2 | XP_006243195.1 | 1 - 2 | 88 | 57 | ||
| GenBank mRNA | FQ213793.1 | - | 1 - 2 | 37 | 57 | |
Set Membership: |
|