Search
Search

RGD1562532| Gene Symbol: RGD1562532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238487.2 | XP_006238549.1 | 3 - 4 | 300 | 119 | |
| XM_008763920.1 | XP_008762142.1 | 2 - 3 | 747 | 119 | ||
| XM_575875.6 | XP_575875.4 | 3 - 4 | 838 | 119 | ||
| GenBank mRNA | BC079057.1 | - | 3 - 4 | 561 | 119 | |
| BC105617.1 | - | 1 - 2 | 162 | 119 | ||
Set Membership: |
|