Search
Search
| Gene Symbol: Inadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238423.2 | XP_006238485.1 | 40 - 41 | 5311 | 155 | |
| XM_006238424.2 | XP_006238486.1 | 40 - 41 | 5317 | 155 | ||
| XM_006238425.2 | XP_006238487.1 | 15 - 16 | 2353 | 155 | ||
| XM_008763916.1 | XP_008762138.1 | 16 - 17 | 2407 | 155 | ||
Set Membership: |
|