Search
Search
| Gene Symbol: Efemp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012039.1 | NP_001012039.1 | 6 - 7 | 735 | 115 | |
| XM_008770456.1 | XP_008768678.1 | 6 - 7 | 835 | 115 | ||
| XM_008770457.1 | XP_008768679.1 | 5 - 6 | 755 | 115 | ||
| GenBank mRNA | BC079455.1 | - | 6 - 7 | 735 | 7121509 | 115 |
| D89730.1 | - | 4 - 5 | 631 | 115 | ||
Set Membership: |
|