Search
Search
| Gene Symbol: Ptgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013015.2 | NP_037147.1 | 5 - 6 | 581 | 146 | |
| GenBank mRNA | FQ214359.1 | - | 4 - 5 | 833 | 146 | |
| J04488.1 | - | 5 - 6 | 581 | 146 | ||
Set Membership: |
|