Search
Search

LOC499746| Gene Symbol: LOC499746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233649.2 | XP_006233711.1 | 2 - 3 | 289 | 127 | |
| XM_006233650.2 | XP_006233712.1 | 2 - 3 | 289 | 127 | ||
| XM_575081.6 | XP_575081.1 | 2 - 3 | 289 | 127 | ||
| GenBank mRNA | BC082090.1 | - | 2 - 3 | 231 | 127 | |
| BC100078.1 | - | 2 - 3 | 240 | 127 | ||
| BC160838.1 | - | 604 | 127 | |||
Set Membership: |
|