Search
Search
| Gene Symbol: Ccdc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766709.1 | XP_008764931.1 | 10 - 11 | 1484 | 126 | |
| XM_008766710.1 | XP_008764932.1 | 10 - 11 | 1481 | 126 | ||
Set Membership: |
|