Search
Search
| Gene Symbol: Pacrgl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006251057.2 | XP_006251119.1 | 3 - 4 | 460 | 96 | |
| XM_008770221.1 | XP_008768443.1 | 3 - 4 | 1268 | 96 | ||
| XM_008770222.1 | XP_008768444.1 | 3 - 4 | 889 | 96 | ||
| XM_341225.7 | XP_341226.5 | 3 - 4 | 369 | 96 | ||
Set Membership: |
|