Search
Search
| Gene Symbol: Acvr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024486.1 | NP_077812.1 | 4 - 5 | 692 | 70 | |
| XM_006234214.2 | XP_006234276.1 | 6 - 7 | 2297 | 70 | ||
| XM_008761862.1 | XP_008760084.1 | 7 - 8 | 2354 | 70 | ||
| XM_008761863.1 | XP_008760085.1 | 7 - 8 | 4598 | 70 | ||
| XM_008761864.1 | XP_008760086.1 | 7 - 8 | 4344 | 70 | ||
| XM_008761865.1 | XP_008760087.1 | 7 - 8 | 4387 | 70 | ||
| XM_008761866.1 | XP_008760088.1 | 7 - 8 | 1064 | 70 | ||
| XM_008761867.1 | XP_008760089.1 | 4 - 5 | 608 | 70 | ||
| GenBank mRNA | BC167754.1 | - | 5 - 6 | 863 | 9089038 | 70 |
| L19341.1 | - | 4 - 5 | 692 | 70 | ||
Set Membership: |
|