Search
Search

Slco1c1| Gene Symbol: Slco1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053441.1 | NP_445893.1 | 8 - 9 | 1409 | 194 | |
| XM_006237602.1 | XP_006237664.1 | 9 - 10 | 1557 | 194 | ||
| GenBank mRNA | AF306546.1 | - | 8 - 9 | 1409 | 194 | |
Set Membership: |
|