Search
Search

Mug1
LOC100911784| Gene Symbol: Mug1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023103.1 | NP_075591.1 | 16 - 17 | 2065 | 177 | |
| XM_006237282.2 | XP_006237344.1 | 16 - 17 | 2105 | 177 | ||
| GenBank mRNA | M22359.1 | - | 935 | 177 | ||
| X52984.1 | - | 16 - 17 | 2065 | 177 | ||
| Gene Symbol: LOC100911784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763278.1 | XP_008761500.1 | 14 - 15 | 1777 | 177 | |
Set Membership: |
|