Search
Search

Kpna2
LOC100359600| Gene Symbol: Kpna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270802.1 | NP_001257731.1 | 2 - 3 | 207 | 117 | |
| NM_053483.2 | NP_445935.2 | 2 - 3 | 182 | 117 | ||
| GenBank mRNA | AJ130946.1 | - | 1 - 2 | 86 | 117 | |
| AY779026.1 | - | 1 - 2 | 76 | 117 | ||
| BC062026.1 | - | 2 - 3 | 182 | 5621957 | 117 | |
| BC089787.1 | - | 2 - 3 | 133 | 7326072 | 117 | |
| FM083931.1 | - | 2 - 3 | 182 | 117 | ||
| FM115539.1 | - | 2 - 3 | 204 | 117 | ||
| Gene Symbol: LOC100359600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729596.4 | XP_002729642.1 | 1 | 202 | 117 | |
Set Membership: |
|