Search
Search

LOC687508
LOC100910772| Gene Symbol: LOC687508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748835.3 | XP_003748883.1 | 3 - 4 | 233 | 83 | |
| XM_008759262.1 | XP_008757484.1 | 4 - 5 | 271 | 83 | ||
| Gene Symbol: LOC100910772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003748816.3 | XP_003748864.1 | 3 - 4 | 652 | 83 | |
Set Membership: |
|