Search
Search
| Gene Symbol: Ms4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107578.1 | NP_001101048.1 | 2 - 3 | 300 | 81 | |
| XM_006231089.2 | XP_006231151.1 | 3 - 4 | 469 | 81 | ||
| GenBank mRNA | BC098745.1 | - | 3 - 4 | 360 | 81 | |
| FQ225563.1 | - | 3 - 4 | 420 | 81 | ||
Set Membership: |
|