Search
Search
| Gene Symbol: Akr1c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138510.1 | NP_612519.1 | 7 - 8 | 889 | 90 | |
| GenBank mRNA | BC086579.1 | - | 7 - 8 | 888 | 90 | |
| BC100248.1 | - | 7 - 8 | 883 | 7307214 | 90 | |
| D14424.1 | - | 7 - 8 | 874 | 90 | ||
| L32601.1 | - | 7 - 8 | 889 | 90 | ||
Set Membership: |
|