Search
Search
| Gene Symbol: Mtx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749167.3 | XP_003749215.1 | 6 - 7 | 617 | 73 | |
| XM_006231788.2 | XP_006231850.1 | 6 - 7 | 617 | 73 | ||
| XM_008760655.1 | XP_008758877.1 | 6 - 7 | 617 | 73 | ||
Set Membership: |
|