Search
Search
| Gene Symbol: Scaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037347.2 | NP_001032424.2 | 7 - 8 | 1101 | 83 | |
| XM_006248024.2 | XP_006248086.1 | 7 - 8 | 1203 | 83 | ||
| XM_006248025.2 | XP_006248087.1 | 7 - 8 | 1203 | 83 | ||
| XM_008768558.1 | XP_008766780.1 | 7 - 8 | 1203 | 83 | ||
| GenBank mRNA | U49058.1 | - | 6 - 7 | 976 | 83 | |
Set Membership: |
|