Search
Search
| Gene Symbol: Cpa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013083.2 | NP_001013101.2 | 6 - 7 | 581 | 80 | |
| GenBank mRNA | CF249031.1 | - | 3 - 4 | 267 | 80 | |
| FQ226028.1 | - | 3 - 4 | 264 | 80 | ||
| FQ234275.1 | - | 2 - 3 | 130 | 80 | ||
Set Membership: |
|