Search
Search
| Gene Symbol: Kifc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103352.1 | NP_001096822.1 | 10 - 11 | 1808 | 62 | |
| XM_006255086.2 | XP_006255148.1 | 14 - 15 | 2110 | 62 | ||
| XM_006255087.2 | XP_006255149.1 | 14 - 15 | 2110 | 62 | ||
| XM_006255088.2 | XP_006255150.1 | 14 - 15 | 2110 | 62 | ||
| XM_008772286.1 | XP_008770508.1 | 14 - 15 | 2107 | 62 | ||
| XM_008772287.1 | XP_008770509.1 | 12 - 13 | 1789 | 62 | ||
| XM_008772288.1 | XP_008770510.1 | 14 - 15 | 2100 | 62 | ||
| XM_008772289.1 | XP_008770511.1 | 13 - 14 | 1883 | 62 | ||
| XM_008772290.1 | XP_008770512.1 | 12 - 13 | 1972 | 62 | ||
| XM_008772291.1 | XP_008770513.1 | 13 - 14 | 1950 | 62 | ||
| XM_008772292.1 | XP_008770514.1 | 11 - 12 | 1579 | 62 | ||
| XM_008772293.1 | XP_008770515.1 | 10 - 11 | 1586 | 62 | ||
| XM_008772294.1 | XP_008770516.1 | 11 - 12 | 3745 | 62 | ||
| XM_008772295.1 | XP_008770517.1 | 12 - 13 | 1778 | 62 | ||
| XM_008772296.1 | XP_008770518.1 | 10 - 11 | 3601 | 62 | ||
| GenBank mRNA | AF035953.1 | - | 1 - 2 | 62 | ||
| AM180764.1 | - | 1 - 2 | 49 | 62 | ||
| BC128748.1 | - | 10 - 11 | 1808 | 7099524 | 62 | |
Set Membership: |
|