Search
Search
| Gene Symbol: Lix1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024303.1 | NP_001019474.1 | 5 - 6 | 774 | 70 | |
| XM_006232978.2 | XP_006233040.1 | 4 - 5 | 794 | 70 | ||
| GenBank mRNA | BC087077.1 | - | 5 - 6 | 774 | 7135157 | 70 |
Set Membership: |
|