Search
Search

Tnfrsf19| Gene Symbol: Tnfrsf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044229.2 | NP_001037694.1 | 3 - 4 | 357 | 81 | |
| GenBank mRNA | DQ466087.1 | - | 3 - 4 | 357 | 81 | |
Set Membership: |
|