Search
Search
| Gene Symbol: Srr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198757.2 | NP_942052.1 | 3 - 4 | 495 | 133 | |
| XM_008768010.1 | XP_008766232.1 | 2 - 3 | 410 | 133 | ||
| XM_008768011.1 | XP_008766233.1 | 2 - 3 | 408 | 133 | ||
| XM_008768012.1 | XP_008766234.1 | 2 - 3 | 291 | 133 | ||
| XM_008768013.1 | XP_008766235.1 | 2 - 3 | 291 | 133 | ||
| XM_008768014.1 | XP_008766236.1 | 2 - 3 | 245 | 133 | ||
| XM_008768015.1 | XP_008766237.1 | 2 - 3 | 313 | 133 | ||
| GenBank mRNA | AB106282.1 | - | 2 - 3 | 265 | 133 | |
| BC082014.1 | - | 3 - 4 | 495 | 7132082 | 133 | |
Set Membership: |
|