Search
Search
| Gene Symbol: C8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191759.1 | NP_001178688.1 | 9 - 10 | 1396 | 80 | |
| XM_006238495.2 | XP_006238557.1 | 10 - 11 | 1716 | 80 | ||
| GenBank mRNA | BC090023.1 | - | 6 - 7 | 921 | 80 | |
| FQ211372.1 | - | 4 - 5 | 380 | 80 | ||
Set Membership: |
|