Search
Search
| Gene Symbol: Gng10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053660.2 | NP_446112.1 | 2 - 3 | 248 | 101 | |
| GenBank mRNA | BC105759.1 | - | 2 - 3 | 241 | 5623275 | 101 |
| BF556593.1 | - | 2 - 3 | 158 | 101 | ||
| CA340326.1 | - | 2 - 3 | 245 | 101 | ||
| CB616034.1 | - | 2 - 3 | 248 | 101 | ||
| DQ120499.1 | - | 1 - 2 | 124 | 101 | ||
| DQ120500.1 | - | 1 - 2 | 124 | 101 | ||
| FQ214425.1 | - | 2 - 3 | 232 | 101 | ||
| FQ229317.1 | - | 2 - 3 | 211 | 101 | ||
| FQ230101.1 | - | 2 - 3 | 211 | 101 | ||
Set Membership: |
|