Search
Search
| Gene Symbol: Dusp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053883.2 | NP_446335.1 | 3 | 2535 | 106 | |
| GenBank mRNA | BC087003.1 | - | 3 | 2535 | 7124105 | 106 |
| FQ217285.1 | - | 2 | 2103 | 107 | ||
Set Membership: |
|