Search
Search
| Gene Symbol: Wdfy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008331.1 | NP_001008332.1 | 4 - 5 | 470 | 70 | |
| XM_006245190.2 | XP_006245252.1 | 5 - 6 | 554 | 70 | ||
| GenBank mRNA | BC085807.1 | - | 4 - 5 | 470 | 7112886 | 70 |
Set Membership: |
|