Search
Search
| Gene Symbol: NFIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005660096.3 | XP_005660153.1 | 1720 | 99 | ||
| XM_021063648.1 | XP_020919307.1 | 1720 | 99 | |||
| XM_021063652.1 | XP_020919311.1 | 5087 | 99 | |||
| XM_021063654.1 | XP_020919313.1 | 1719 | 99 | |||
| XM_021063657.1 | XP_020919316.1 | 1718 | 99 | |||
| XM_021063662.1 | XP_020919321.1 | 1718 | 99 | |||
| XM_021063669.1 | XP_020919328.1 | 1716 | 99 | |||
| XM_021063671.1 | XP_020919330.1 | 1717 | 99 | |||
| XM_021063676.1 | XP_020919335.1 | 1713 | 99 | |||
| XM_021063685.1 | XP_020919344.1 | 5087 | 99 | |||
| GenBank mRNA | AK236442.1 | - | 2 - 3 | 1562 | 99 | |
| AK393433.1 | - | 2 - 3 | 1615 | 99 | ||
| AK395543.1 | - | 2 - 3 | 1079 | 99 | ||
| AK400858.1 | - | 2 - 3 | 1176 | 99 | ||
| AY609566.1 | - | 2 - 3 | 606 | 99 | ||
Set Membership: |
|