Search
Search
| Gene Symbol: NTRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064642.1 | XP_020920301.1 | 2029 | 134 | ||
| XM_021064643.1 | XP_020920302.1 | 1906 | 134 | |||
| XM_021064644.1 | XP_020920303.1 | 2104 | 134 | |||
| XM_021064645.1 | XP_020920304.1 | 2007 | 134 | |||
| XM_021064646.1 | XP_020920305.1 | 2069 | 134 | |||
| XM_021064647.1 | XP_020920306.1 | 1910 | 134 | |||
| XM_021064648.1 | XP_020920307.1 | 2017 | 134 | |||
| XM_021064649.1 | XP_020920308.1 | 2016 | 134 | |||
| XM_021064650.1 | XP_020920309.1 | 2011 | 134 | |||
| GenBank mRNA | AJ278437.1 | - | 1 | 154 | 134 | |
| AK231926.1 | - | 2 | 299 | 134 | ||
| AK345762.1 | - | 2 | 282 | 134 | ||
| AK390000.1 | - | 10 | 1307 | 134 | ||
| AK390259.1 | - | 9 | 1756 | 134 | ||
| AK393897.1 | - | 9 | 1660 | 134 | ||
| AK397968.1 | - | 10 | 1854 | 134 | ||
| AK400263.1 | - | 10 | 1900 | 134 | ||
| AK400521.1 | - | 9 | 1677 | 134 | ||
Set Membership: |
|