Search
Search
| Gene Symbol: STC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103212.1 | NP_001096682.1 | 1 - 2 | 140 | 78 | |
| GenBank mRNA | AK234059.1 | - | 1 - 2 | 341 | 78 | |
| EU086592.1 | - | 1 - 2 | 140 | 78 | ||
Set Membership: |
|