Search
Search

ANGPTL1| Gene Symbol: ANGPTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109947.1 | NP_001103417.1 | 2 - 3 | 1022 | 141 | |
| XM_005667787.3 | XP_005667844.1 | 5269 | 141 | |||
| GenBank mRNA | EF154830.1 | - | 2 - 3 | 1022 | 141 | |
Set Membership: |
|