Search
Search
| Gene Symbol: KDM1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112687.1 | NP_001106157.1 | 5 - 6 | 846 | 72 | |
| XM_021093198.1 | XP_020948857.1 | 935 | 72 | |||
| XM_021093199.1 | XP_020948858.1 | 875 | 72 | |||
| XM_021093200.1 | XP_020948859.1 | 874 | 72 | |||
| GenBank mRNA | AK349640.1 | - | 2 - 3 | 131 | 72 | |
| EU219915.1 | - | 5 - 6 | 846 | 72 | ||
| EU219916.1 | - | 4 - 5 | 786 | 72 | ||
Set Membership: |
|