Search
Search

CYP2D25| Gene Symbol: CYP2D25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214394.1 | NP_999559.1 | 8 - 9 | 1394 | 135 | |
| GenBank mRNA | AK232448.1 | - | 8 - 9 | 1410 | 135 | |
| AK232749.1 | - | 8 - 9 | 1402 | 135 | ||
| AK394110.1 | - | 7 - 8 | 1003 | 135 | ||
| AK398955.1 | - | 8 - 9 | 1400 | 135 | ||
| AK400510.1 | - | 8 - 9 | 1400 | 135 | ||
| D89502.1 | - | 8 - 9 | 1359 | 135 | ||
| KP687264.1 | - | 8 - 9 | 1508 | 135 | ||
| Y16417.1 | - | 1394 | 135 | |||
Set Membership: |
|