Search
Search
| Gene Symbol: ACTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134354.1 | NP_001127826.1 | 8 - 9 | 1009 | 91 | |
| XM_005662508.3 | XP_005662565.1 | 1196 | 91 | |||
| GenBank mRNA | EU868831.1 | - | 8 - 9 | 1009 | 91 | |
Set Membership: |
|