Search
Search
| Gene Symbol: NLRX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204769.1 | NP_001191698.1 | 6 - 7 | 2287 | 61 | |
| XM_005667404.3 | XP_005667461.1 | 61 | ||||
| XM_005667405.3 | XP_005667462.1 | 61 | ||||
| XM_021102056.1 | XP_020957715.1 | 61 | ||||
| XM_021102057.1 | XP_020957716.1 | 61 | ||||
| XM_021102058.1 | XP_020957717.1 | 61 | ||||
| GenBank mRNA | GU989323.1 | - | 6 - 7 | 2287 | 61 | |
Set Membership: |
|