Search
Search
| Gene Symbol: TNR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013989629.2 | XP_013845083.1 | 1952 | 97 | ||
| XM_021063665.1 | XP_020919324.1 | 2171 | 97 | |||
| XM_021063666.1 | XP_020919325.1 | 1922 | 97 | |||
| XM_021063667.1 | XP_020919326.1 | 9931 | 97 | |||
| XM_021063668.1 | XP_020919327.1 | 362 | 97 | |||
Set Membership: |
|