Search
Search
| Gene Symbol: CA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067239.1 | XP_020922898.1 | 1922 | 80 | ||
| XM_021067240.1 | XP_020922899.1 | 2638 | 80 | |||
| XM_021067241.1 | XP_020922900.1 | 1625 | 80 | |||
| XM_021067242.1 | XP_020922901.1 | 1112 | 80 | |||