Search
Search

FAM135B| Gene Symbol: FAM135B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088864.1 | XP_020944523.1 | 4019 | 70 | ||
| XM_021088865.1 | XP_020944524.1 | 4642 | 70 | |||
| XM_021088866.1 | XP_020944525.1 | 3969 | 70 | |||
| XM_021088867.1 | XP_020944526.1 | 3543 | 70 | |||
| XM_021088868.1 | XP_020944527.1 | 3916 | 70 | |||
| XM_021088869.1 | XP_020944528.1 | 4562 | 70 | |||
| XM_021088870.1 | XP_020944529.1 | 3884 | 70 | |||