Search
Search
| Gene Symbol: PTPRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926867.7 | XP_001926902.5 | 4998 | 107 | ||
| XM_021091987.1 | XP_020947646.1 | 5708 | 107 | |||
| XM_021091988.1 | XP_020947647.1 | 5705 | 107 | |||
| XM_021091989.1 | XP_020947648.1 | 5444 | 107 | |||
Set Membership: |
|