Search
Search
| Gene Symbol: KLHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123968.5 | XP_003124016.2 | 987 | 104 | ||
| XM_003123969.4 | XP_003124017.1 | 1224 | 104 | |||
| XM_005661680.3 | XP_005661737.1 | 946 | 104 | |||
| XM_013995146.2 | XP_013850600.1 | 1189 | 104 | |||
| XM_013995147.2 | XP_013850601.1 | 881 | 104 | |||
| XM_013995148.2 | XP_013850602.1 | 644 | 104 | |||
| XM_021084789.1 | XP_020940448.1 | 637 | 104 | |||
| XM_021084790.1 | XP_020940449.1 | 534 | 104 | |||
| XM_021084791.1 | XP_020940450.1 | 591 | 104 | |||
| XM_021084792.1 | XP_020940451.1 | 1123 | 104 | |||
| XM_021084793.1 | XP_020940452.1 | 845 | 104 | |||
| XM_021084794.1 | XP_020940453.1 | 537 | 104 | |||
| XM_021084795.1 | XP_020940454.1 | 1128 | 104 | |||
| XM_021084796.1 | XP_020940455.1 | 1799 | 104 | |||
| XM_021084797.1 | XP_020940456.1 | 231 | 104 | |||
| XM_021084798.1 | XP_020940457.1 | 1867 | 104 | |||
| GenBank mRNA | AK233262.1 | - | 5 - 6 | 878 | 104 | |
Set Membership: |
|