Search
Search
| Gene Symbol: BRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081614.1 | XP_020937273.1 | 3012 | 51 | ||
| XM_021081615.1 | XP_020937274.1 | 2928 | 51 | |||
| XM_021081616.1 | XP_020937275.1 | 1632 | 51 | |||
| XM_021081617.1 | XP_020937276.1 | 1527 | 51 | |||
| XM_021081618.1 | XP_020937277.1 | 3430 | 51 | |||
Set Membership: |
|